Assay Details
Target Gene Details
Entrez Gene ID: | 3508 |
Gene Name: | immunoglobulin mu binding protein 2 |
Gene Aliases: |
CATF1, CMT2S, HCSA, HMN6, SMARD1, SMUBP2, ZFAND7 |
Location: |
Chr.11:68903851-68940602 on Build GRCh38 |
Assay Gene Location: | Within Exon 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| IGHMBP2 | NM_002180.2 | 13 | 2370 | NP_002171.2 |
| XM_005273974.2 | 13 | 2334 | XP_005274031.1 | |
| XM_005273975.3 | 6 | 1465 | XP_005274032.1 | |
| XM_011544994.1 | 6 | 1852 | XP_011543296.1 | |
| XM_017017669.1 | 12 | 2226 | XP_016873158.1 | |
| XM_017017670.1 | 9 | 2131 | XP_016873159.1 | |
| AK091539.1 | 4 | 1261 | ||
| AK123166.1 | 5 | 2027 | ||
| AK312565.1 | 13 | 2328 | ||
| BC000290.2 | 13 | 2283 | AAH00290.1 | |
| BC025299.1 | 13 | 2315 | ||
| BC105088.1 | 13 | 2307 | AAI05089.1 | |
| BC105090.1 | 13 | 2307 | AAI05091.1 | |
| L14754.1 | 13 | 2317 | AAA53082.1 | |
| M64979.1 | 4 | 798 | AAA58611.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv509423 | Chr.11:68873995 - 68970585 on Build GRCh38 | Insertion |
|
| nsv1046306 | Chr.11:68933054 - 68969593 on Build GRCh38 | Gain |
|
| nsv428261 | Chr.11:68754616 - 68937835 on Build GRCh38 | Gain |
|
| esv3579736 | Chr.11:68929224 - 68943379 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map