Assay Details
Target Gene Details
Entrez Gene ID: | 219541 |
Gene Name: | mediator complex subunit 19 |
Gene Aliases: |
DT2P1G7, LCMR1, MED19AS |
Location: |
Chr.11:57703709-57712323 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MED19 | NM_001317078.1 | 5 | 947 | NP_001304007.1 |
| NM_153450.2 | 4 | 1381 | NP_703151.2 | |
| AA972146.1 | ||||
| AY148462.1 | 5 | 676 | AAN16075.1 | |
| BC037223.1 | 4 | 1259 | AAH37223.1 | |
| EB386171.1 | ||||
| U92995.1 | 4 | 381 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3579728 | Chr.11:57686071 - 57726599 on Build GRCh38 | Loss |
|
| nsv832163 | Chr.11:57531881 - 57713578 on Build GRCh38 | Gain |
|
| nsv508633 | Chr.11:57652112 - 57792573 on Build GRCh38 | Deletion |
|
| nsv832164 | Chr.11:57640144 - 57811353 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map