Assay Details
Target Gene Details
Entrez Gene ID: | 10457 |
Gene Name: | glycoprotein nmb |
Gene Aliases: |
HGFIN, NMB |
Location: |
Chr.7:23246206-23275110 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GPNMB | NM_001005340.1 | 2 | 281 | NP_001005340.1 |
| NM_002510.2 | 2 | 281 | NP_002501.1 | |
| XM_005249578.2 | 2 | 772 | XP_005249635.1 | |
| XM_017011676.1 | 2 | 772 | XP_016867165.1 | |
| XM_017011677.1 | 2 | 772 | XP_016867166.1 | |
| XM_017011678.1 | 2 | 772 | XP_016867167.1 | |
| AF322909.1 | 2 | 179 | AAG42839.1 | |
| AJ505015.1 | 2 | 193 | CAD43718.1 | |
| AK292078.1 | 2 | 203 | ||
| AK296779.1 | 2 | 203 | ||
| AK297031.1 | 2 | 203 | ||
| AK297051.1 | 2 | 212 | ||
| AK297055.1 | ||||
| AK297803.1 | ||||
| AK303071.1 | ||||
| AK304671.1 | 3 | 262 | ||
| AK312279.1 | 2 | 210 | ||
| AY359124.1 | 2 | 201 | AAQ89481.1 | |
| BC011595.1 | 2 | 169 | AAH11595.1 | |
| BC032783.1 | 2 | 197 | AAH32783.1 | |
| BM750843.1 | 2 | 291 | ||
| BT007074.1 | 2 | 120 | AAP35737.1 | |
| CD105306.1 | 2 | 269 | ||
| X76534.1 | 2 | 211 | CAA54044.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1027363 | Chr.7:22817000 - 23271413 on Build GRCh38 | Gain |
|
| nsv1016316 | Chr.7:23221900 - 23318018 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map