Assay Details
Target Gene Details
Entrez Gene ID: | 283365 |
Gene Name: | olfactory receptor family 6 subfamily C member 6 |
Gene Aliases: |
- |
Location: |
Chr.12:55294288-55295232 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| OR6C6 | NM_001005493.1 | 1 | 437 | NP_001005493.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv559001 | Chr.12:55113681 - 55663924 on Build GRCh38 | Loss |
|
| nsv826381 | Chr.12:55276757 - 55331791 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map