Assay Details
Target Gene Details
Entrez Gene ID: | 54431 |
Gene Name: | DnaJ heat shock protein family (Hsp40) member C10 |
Gene Aliases: |
ERdj5, JPDI, MTHr, PDIA19 |
Location: |
Chr.2:182716041-182780025 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DNAJC10 | NM_001271581.1 | 1 | 339 | NP_001258510.1 |
| NM_018981.2 | 1 | 339 | NP_061854.1 | |
| NR_073365.1 | 1 | 339 | ||
| NR_073366.1 | 1 | 339 | ||
| NR_073367.1 | 1 | 339 | ||
| AF038503.1 | AAN73271.1 | |||
| AF490904.1 | 1 | 87 | AAM09527.1 | |
| AK027647.1 | 1 | 108 | BAB55263.1 | |
| AK027696.1 | 1 | 87 | BAB55304.1 | |
| AL832632.2 | 1 | 110 | ||
| AY358577.1 | 1 | 96 | AAQ88940.1 | |
| BC117299.1 | 1 | 54 | ||
| BC126168.1 | 1 | 54 | ||
| DA106532.1 | 1 | 339 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv34847 | Chr.2:182672672 - 182922035 on Build GRCh38 | Gain |
|
| esv2759104 | Chr.2:182572973 - 182970356 on Build GRCh38 | Gain |
|
| nsv1009882 | Chr.2:182348425 - 183306791 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map