Assay Details
Target Gene Details
Entrez Gene ID: | 56260 |
Gene Name: | chromosome 8 open reading frame 44 |
Gene Aliases: |
- |
Location: |
Chr.8:66667552-66681149 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| C8orf44 | NM_019607.2 | 3 | 1392 | NP_062553.1 |
| AA551208.1 | ||||
| AK002129.1 | 4 | 1466 | BAA92096.1 | |
| BC014448.1 | 3 | 1326 | AAH14448.1 |
Target Gene Details
Entrez Gene ID: | 100533105 |
Gene Name: | C8orf44-SGK3 readthrough |
Gene Aliases: |
- |
Location: |
Chr.8:66667552-66862022 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| C8orf44-SGK3 | NM_001204173.1 | NP_001191102.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv831346 | Chr.8:66574320 - 66763089 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map