Assay Details
Target Gene Details
Entrez Gene ID: | 101926905 |
Gene Name: | uncharacterized LOC101926905 |
Gene Aliases: |
- |
Location: |
Chr.5:141046205-141078031 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC101926905 | NR_105056.2 | |||
| BG193185.1 | ||||
| HY003594.1 | ||||
| HY010779.1 |
Target Gene Details
Entrez Gene ID: | 29930 |
Gene Name: | protocadherin beta 1 |
Gene Aliases: |
PCDH-BETA1 |
Location: |
Chr.5:141051376-141053962 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PCDHB1 | NM_013340.3 | 1 | 1151 | NP_037472.2 |
| AF152488.1 | 1 | 1056 | AAD43749.1 | |
| BC112096.1 | 1 | 1151 | AAI12097.1 | |
| BC112098.1 | 1 | 1151 | AAI12099.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2759382 | Chr.5:140777355 - 141449115 on Build GRCh38 | Gain |
|
| nsv428127 | Chr.5:141016768 - 141319173 on Build GRCh38 | Loss |
|
| nsv818387 | Chr.5:141040406 - 141056478 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map