Assay Details
Target Gene Details
Entrez Gene ID: | 10289 |
Gene Name: | eukaryotic translation initiation factor 1B |
Gene Aliases: |
GC20 |
Location: |
Chr.3:40309682-40312424 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| EIF1B | NM_005875.2 | 1 | 185 | NP_005866.1 |
| AF064607.1 | AAC17112.1 | |||
| AF077052.1 | 1 | 167 | AAD27785.1 | |
| AF263452.1 | 1 | 159 | AAF79182.1 | |
| AK223448.1 | 1 | 180 | BAD97168.1 | |
| AK311922.1 | 1 | 88 | ||
| BC006996.1 | 1 | 169 | AAH06996.1 | |
| BF667107.1 | 1 | 184 | ||
| CA335728.1 | 1 | 198 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv834667 | Chr.3:40149053 - 40317061 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map