Assay Details
Target Gene Details
Entrez Gene ID: | 26354 |
Gene Name: | G protein nucleolar 3 |
Gene Aliases: |
C77032, E2IG3, NNP47, NS |
Location: |
Chr.3:52685920-52694497 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GNL3 | NM_014366.4 | 1 | 71 | NP_055181.3 |
| NM_206825.1 | 1 | 71 | NP_996561.1 | |
| NM_206826.1 | 1 | 71 | NP_996562.1 | |
| AF191018.1 | 1 | 71 | AAF09482.1 | |
| AI028402.1 | 1 | 71 | ||
| AK027514.1 | BAB55168.1 | |||
| AK315484.1 | ||||
| BF732788.1 | 1 | 71 |
More Information
Additional Information:
For this assay, SNP(s) [rs78053665] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map