Assay Details
Target Gene Details
Entrez Gene ID: | 22883 |
Gene Name: | calsyntenin 1 |
Gene Aliases: |
ALC-ALPHA, CDHR12, CST-1, CSTN1, PIK3CD, XB31alpha, alcalpha1, alcalpha2 |
Location: |
Chr.1:9729021-9824526 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CLSTN1 | NM_001009566.2 | 10 | 2266 | NP_001009566.1 |
| NM_001302883.1 | 10 | 2266 | NP_001289812.1 | |
| NM_014944.4 | 9 | 2236 | NP_055759.3 | |
| AB020718.1 | 10 | 2266 | ||
| AF438482.1 | 9 | 2202 | AAQ04552.1 | |
| AK000605.1 | 6 | 933 | ||
| AK289798.1 | 9 | 1444 | ||
| AK304813.1 | 10 | 1508 | ||
| AY753301.1 | 9 | 1443 | AAV30551.1 | |
| BC033902.2 | 9 | 1694 | AAH33902.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832203 | Chr.1:9666574 - 9800151 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map