Assay Details
Target Gene Details
Entrez Gene ID: | 84986 |
Gene Name: | Rho GTPase activating protein 19 |
Gene Aliases: |
- |
Location: |
Chr.10:97222173-97292673 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 11 - Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ARHGAP19 | NM_001204300.1 | NP_001191229.1 | ||
| NM_001256423.1 | NP_001243352.1 | |||
| NM_032900.5 | NP_116289.4 | |||
| AK074122.1 | BAB84948.1 | |||
| AK090447.1 | BAC03428.1 | |||
| AK093316.1 | BAC04130.1 | |||
| AK093441.1 | BAC04165.1 | |||
| AK303055.1 | ||||
| BC113888.1 | ||||
| BC114490.1 | ||||
| BX648189.1 | ||||
| BX648194.1 | ||||
| CR749412.1 | CAH18254.1 | |||
| DQ338460.1 |
Target Gene Details
Entrez Gene ID: | 100533184 |
Gene Name: | ARHGAP19-SLIT1 readthrough (NMD candidate) |
Gene Aliases: |
- |
Location: |
Chr.10:97153042-97292673 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 10 - Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ARHGAP19-SLIT1 | NR_037909.1 | |||
| AY336750.1 | AAR02412.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1040903 | Chr.10:97226625 - 97287718 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map