Assay Details
Target Gene Details
Entrez Gene ID: | 64780 |
Gene Name: | microtubule associated monooxygenase, calponin and LIM domain containing 1 |
Gene Aliases: |
MICAL, MICAL-1, NICAL |
Location: |
Chr.6:109444062-109465968 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 26 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MICAL1 | NM_001159291.1 | NP_001152763.1 | ||
| NM_001286613.1 | NP_001273542.1 | |||
| NM_022765.3 | NP_073602.3 | |||
| AB048948.1 | BAB86289.1 | |||
| AK021999.1 | BAB13949.1 | |||
| AK024500.1 | BAB15790.1 | |||
| AK025392.1 | BAB15124.1 | |||
| AK160384.1 | BAD18727.1 | |||
| AK225049.1 | ||||
| AK296284.1 | ||||
| AL122098.1 | CAB59266.1 | |||
| BC009972.2 | AAH09972.2 | |||
| BC042144.1 | AAH42144.1 | |||
| BC052983.1 | AAH52983.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3576210 | Chr.6:109428085 - 109446498 on Build GRCh38 | Gain |
|
| nsv950493 | Chr.6:109442398 - 109456197 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map