Assay Details
Target Gene Details
Entrez Gene ID: | 121256 |
Gene Name: | transmembrane protein 132D |
Gene Aliases: |
MOLT, PPP1R153 |
Location: |
Chr.12:129071726-129903667 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TMEM132D | NM_133448.2 | 9 | 4189 | NP_597705.2 |
| AB061814.1 | 9 | 4190 | BAC97794.1 | |
| AB075824.1 | 4 | 3716 | BAB85530.1 | |
| AK094656.1 | 1 | 1023 | ||
| AL833234.1 | 1 | 1312 | CAD38636.1 | |
| BC114626.1 | ||||
| BC114936.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv930 | Chr.12:129065704 - 129096684 on Build GRCh38 | Deletion |
|
| esv3631156 | Chr.12:128909343 - 129187331 on Build GRCh38 | Gain |
|
| nsv1040831 | Chr.12:128749443 - 129516029 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map