Assay Details
Target Gene Details
Entrez Gene ID: | 51374 |
Gene Name: | all-trans retinoic acid induced differentiation factor |
Gene Aliases: |
APR--3, APR-3, APR3, C2orf28, HSPC013, PRO240, p18 |
Location: |
Chr.2:27212031-27217178 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ATRAID | NM_001170795.1 | 2 | 205 | NP_001164266.1 |
| NM_016085.4 | 2 | 345 | NP_057169.2 | |
| NM_080592.3 | 2 | 525 | NP_542159.3 | |
| AB009017.1 | 2 | 218 | ||
| AF077037.1 | 2 | 207 | AAD27770.1 | |
| AF144055.2 | 2 | 336 | AAD31317.2 | |
| AF275744.1 | 3 | 199 | AAK69412.1 | |
| AK291894.1 | 2 | 335 | ||
| AY358968.1 | 2 | 198 | AAQ89327.1 | |
| BC002846.1 | 2 | 193 | ||
| BC011006.1 | 2 | 199 | ||
| BC021237.2 | 2 | 305 | AAH21237.1 | |
| BC035850.1 | 2 | 191 | AAH35850.1 | |
| BI194121.1 | 2 | 345 | ||
| BQ420456.1 | 2 | 328 | ||
| BQ718371.1 | 2 | 240 | ||
| CA450289.1 | 6 | 730 | ||
| CF126865.1 | 2 | 338 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833603 | Chr.2:27059106 - 27263631 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map