Assay Details
Target Gene Details
Entrez Gene ID: | 6628 |
Gene Name: | small nuclear ribonucleoprotein polypeptides B and B1 |
Gene Aliases: |
CCMS, COD, SNRPB1, Sm-B/B', SmB/B', SmB/SmB', snRNP-B |
Location: |
Chr.20:2461635-2470853 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SNRPB | NM_003091.3 | NP_003082.1 | ||
| NM_198216.1 | NP_937859.1 | |||
| AK301034.1 | ||||
| AK301206.1 | ||||
| AK312620.1 | ||||
| AY742712.1 | AAU89981.1 | |||
| BC080516.1 | AAH80516.1 | |||
| CR456969.1 | CAG33250.1 | |||
| J04564.1 | AAA60151.1 | |||
| M34081.1 | AAA36578.1 | |||
| M34082.1 | AAA36579.1 | |||
| X15893.1 | CAA33902.1 | |||
| X17567.1 | CAB57867.1 | |||
| X17568.1 | CAB57868.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv470535 | Chr.20:1948896 - 2780594 on Build GRCh38 | Gain |
|
| esv3644998 | Chr.20:2148722 - 2690694 on Build GRCh38 | Gain |
|
| esv2758778 | Chr.20:2149914 - 2494823 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map