Assay Details
Target Gene Details
Entrez Gene ID: | 404734 |
Gene Name: | ANKHD1-EIF4EBP3 readthrough |
Gene Aliases: |
MASK-BP3, MASK-BP3ARF |
Location: |
Chr.5:140401814-140549578 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 33 - Exon 34 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ANKHD1-EIF4EBP3 | NM_020690.5 | NP_065741.3 | ||
| AB029008.1 | 5 | 4849 | BAA83037.1 | |
| AF521883.1 | AAO14944.1 | |||
| AK125734.1 |
More Information
Set Membership: |
|

Genomic Map