Assay Details
Target Gene Details
Entrez Gene ID: | 55559 |
Gene Name: | HAUS augmin like complex subunit 7 |
Gene Aliases: |
UCHL5IP, UIP1 |
Location: |
Chr.X:153447665-153495525 on Build GRCh38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HAUS7 | NM_017518.7 | 8 | 883 | NP_059988.3 |
| NR_073156.1 | ||||
| AF267739.1 | 8 | 814 | AAF75785.1 | |
| AK125219.1 | BAC86087.1 | |||
| AK300650.1 | 8 | 835 | ||
| AK302066.1 | ||||
| AL122036.1 | 4 | 418 | CAB59176.1 | |
| BC008141.1 | 8 | 883 | AAH08141.3 | |
| CA441020.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3577532 | Chr.X:153441652 - 153467949 on Build GRCh38 | Gain |
|
| esv33199 | Chr.X:153065734 - 154591978 on Build GRCh38 | Gain+Loss |
|
| esv3575022 | Chr.X:153455009 - 153456267 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map