Assay Details
Target Gene Details
Entrez Gene ID: | 203100 |
Gene Name: | HtrA serine peptidase 4 |
Gene Aliases: |
- |
Location: |
Chr.8:38974149-38988662 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HTRA4 | NM_153692.3 | 1 | 102 | NP_710159.1 |
| XM_011544431.2 | 1 | 102 | XP_011542733.1 | |
| XM_011544432.1 | 1 | 102 | XP_011542734.1 | |
| AK075205.1 | 1 | 73 | BAC11470.1 | |
| AK300197.1 | ||||
| BC057765.1 | AAH57765.1 | |||
| DC423042.1 | 1 | 259 |
More Information
Additional Information:
For this assay, SNP(s) [rs115575935] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map