Assay Details
Target Gene Details
Entrez Gene ID: | 53 |
Gene Name: | acid phosphatase 2, lysosomal |
Gene Aliases: |
LAP |
Location: |
Chr.11:47239302-47248847 on Build GRCh38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ACP2 | NM_001302489.1 | 11 | 1549 | NP_001289418.1 |
| NM_001302490.1 | 10 | 1481 | NP_001289419.1 | |
| NM_001302491.1 | 9 | 1388 | NP_001289420.1 | |
| NM_001302492.1 | 10 | 1506 | NP_001289421.1 | |
| NM_001610.3 | 11 | 1577 | NP_001601.1 | |
| AK298122.1 | 9 | 1343 | ||
| AK301860.1 | ||||
| AK303894.1 | ||||
| AK308512.1 | 9 | 1796 | ||
| AY007137.1 | 8 | 1109 | ||
| BC003160.1 | 11 | 1522 | AAH03160.1 | |
| BC093010.1 | 11 | 1535 | AAH93010.1 | |
| X12548.1 | 11 | 1531 | CAA31064.1 |
More Information
Set Membership: |
|

Genomic Map