Assay Details
Target Gene Details
Entrez Gene ID: | 84720 |
Gene Name: | phosphatidylinositol glycan anchor biosynthesis class O |
Gene Aliases: |
HPMRS2 |
Location: |
Chr.9:35085493-35096601 on Build GRCh38 |
Assay Gene Location: | Within Exon 11 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PIGO | NM_001201484.1 | 13 | 2675 | NP_001188413.1 |
NM_032634.3 | 11 | 3828 | NP_116023.2 | |
NM_152850.3 | 12 | 2223 | NP_690577.2 | |
XM_005251619.3 | 11 | 3453 | XP_005251676.1 | |
XM_017015222.1 | 12 | 3945 | XP_016870711.1 | |
XM_017015223.1 | 12 | 2604 | XP_016870712.1 | |
XM_017015224.1 | 12 | 2868 | XP_016870713.1 | |
AK074064.1 | 5 | 5210 | BAB84890.1 | |
AK090433.1 | 4 | 6415 | BAC03414.1 | |
AL833956.1 | 11 | 3861 | CAD38806.1 | |
AW149348.1 | 1 | 244 | ||
AY358472.1 | 10 | 3434 | AAQ88836.1 | |
BC001030.2 | 9 | 1553 | AAH01030.1 | |
BC013987.2 | 10 | 1639 | AAH13987.1 | |
BC029271.1 | 10 | 3099 | AAH29271.1 | |
BC036916.1 | 11 | 2188 | AAH36916.1 | |
BC065282.1 | 13 | 2346 | AAH65282.1 | |
BE139632.1 | 1 | 238 | ||
BU727886.1 | 1 | 260 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1030303 | Chr.9:35084357 - 35506646 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
