Assay Details
Target Gene Details
Entrez Gene ID: | 1316 |
Gene Name: | Kruppel like factor 6 |
Gene Aliases: |
BCD1, CBA1, COPEB, CPBP, GBF, PAC1, ST12, ZF9 |
Location: |
Chr.10:3775996-3785281 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KLF6 | NM_001160124.1 | 4 | 1277 | NP_001153596.1 |
| NM_001160125.1 | 3 | 1279 | NP_001153597.1 | |
| NM_001300.5 | 4 | 1403 | NP_001291.3 | |
| NR_027653.1 | 4 | 1249 | ||
| AB017493.1 | 4 | 1253 | BAA33050.1 | |
| AF001461.1 | 4 | 1166 | AAC39929.1 | |
| AI355637.1 | ||||
| BC000311.2 | 4 | 1322 | AAH00311.1 | |
| BX537374.1 | 2 | 1761 | CAD97616.1 | |
| CD742988.1 | ||||
| U44975.1 | 4 | 1157 | AAC23699.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv661n100 | Chr.10:3749938 - 3788015 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map