Assay Details
Target Gene Details
Entrez Gene ID: | 3791 |
Gene Name: | kinase insert domain receptor |
Gene Aliases: |
CD309, FLK1, VEGFR, VEGFR2 |
Location: |
Chr.4:55078259-55125595 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 13 - Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KDR | NM_002253.2 | NP_002244.1 | ||
| AB209901.1 | BAD93138.1 | |||
| AF035121.1 | AAB88005.1 | |||
| AF063658.1 | AAC16450.1 | |||
| BC131822.1 | ||||
| EU826563.1 | ||||
| L04947.1 | AAA59459.1 | |||
| X61656.1 | CAA43837.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv526344 | Chr.4:54656321 - 55364952 on Build GRCh38 | Gain |
|
| nsv829939 | Chr.4:54990604 - 55138844 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map