Assay Details
Target Gene Details
Entrez Gene ID: | 121506 |
Gene Name: | endoplasmic reticulum protein 27 |
Gene Aliases: |
C12orf46, PDIA8 |
Location: |
Chr.12:14914027-14938549 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ERP27 | NM_001300784.1 | NP_001287713.1 | ||
| NM_152321.3 | NP_689534.1 | |||
| AK056677.1 | BAB71251.1 | |||
| AY358536.1 | AAQ88900.1 | |||
| BC030218.1 | AAH30218.1 | |||
| CD389092.1 | ||||
| CN296050.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs79989395] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map