Assay Details
Target Gene Details
Entrez Gene ID: | 51292 |
Gene Name: | guanosine monophosphate reductase 2 |
Gene Aliases: |
GMPR 2 |
Location: |
Chr.14:24232417-24239242 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GMPR2 | NM_001002000.2 | 1 | 238 | NP_001002000.1 |
| NM_001002001.2 | 1 | 238 | NP_001002001.1 | |
| NM_001283021.1 | 1 | 238 | NP_001269950.1 | |
| NR_104265.1 | 1 | 238 | ||
| XM_005267740.3 | 1 | 103 | XP_005267797.1 | |
| XM_005267741.3 | 1 | 103 | XP_005267798.1 | |
| XM_017021356.1 | 1 | 103 | XP_016876845.1 | |
| XM_017021357.1 | 1 | 201 | XP_016876846.1 | |
| XM_017021358.1 | 1 | 243 | XP_016876847.1 | |
| AF086919.1 | AAP97149.1 | |||
| AF135159.1 | 1 | 183 | AAG09132.1 | |
| AF419346.1 | 1 | 29 | AAN32701.1 | |
| AK226166.1 | ||||
| BC093039.1 | 1 | 36 | AAH93039.1 | |
| BE003291.1 | 1 | 236 | ||
| BX161436.1 | 1 | 218 | CAD61908.1 | |
| BX161518.1 | CAD61951.1 |
More Information
Set Membership: |
|

Genomic Map