Assay Details
Target Gene Details
Entrez Gene ID: | 400794 |
Gene Name: | uncharacterized LOC400794 |
Gene Aliases: |
- |
Location: |
Chr.1:165476842-165582155 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC400794 | NR_026744.2 | |||
| AK093132.1 |
Target Gene Details
Entrez Gene ID: | 440699 |
Gene Name: | leucine rich repeat containing 52 |
Gene Aliases: |
- |
Location: |
Chr.1:165544241-165563948 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LRRC52 | NM_001005214.3 | 1 | 269 | NP_001005214.2 |
| AK098677.1 | 1 | 505 | BAC05375.1 | |
| BC132981.1 | 1 | 241 | ||
| BC132983.1 | 1 | 241 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv3376 | Chr.1:165513885 - 165548586 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map