Assay Details
Target Gene Details
Entrez Gene ID: | 2975 |
Gene Name: | general transcription factor IIIC subunit 1 |
Gene Aliases: |
TFIIIC, TFIIIC220, TFIIICalpha |
Location: |
Chr.16:27460613-27549930 on Build GRCh38 |
Assay Gene Location: | Within Exon 36 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GTF3C1 | NM_001286242.1 | 36 | 5952 | NP_001273171.1 |
| NM_001520.3 | 36 | 6027 | NP_001511.2 | |
| XM_017023188.1 | 35 | 5834 | XP_016878677.1 | |
| AK091032.1 | 1 | 1050 | ||
| AK123999.1 | 3 | 1449 | ||
| AK131479.1 | 3 | 2431 | ||
| BC044857.2 | 16 | 2594 | AAH44857.1 | |
| BC052642.1 | 1 | 68 | AAH52642.1 | |
| BC137229.1 | 36 | 6006 | ||
| BX648143.1 | 36 | 5996 | CAH10771.1 | |
| CR627430.1 | 36 | 6012 | CAH10517.1 | |
| U02619.1 | 36 | 6047 | AAA17985.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1059454 | Chr.16:27139937 - 27679877 on Build GRCh38 | Gain |
|
| nsv833179 | Chr.16:27341363 - 27496185 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map