| nsv509709 | 
						
							
								Chr.18:79685368 - 79838375
							    on Build GRCh38
						
							
						 | 
						Insertion | 
						
							
								 LOC105376875
							
								 CTDP1
							
								 KCNG2
							
						 | 
					
				
		    		
						| nsv428359 | 
						
							
								Chr.18:79277548 - 80255221
							    on Build GRCh38
						
							
						 | 
						Gain | 
						
							
								 CTDP1
							
								 LOC284241
							
								 PARD6G-AS1
							
								 PQLC1
							
								 KCNG2
							
								 ADNP2
							
								 RBFADN
							
								 ATP9B
							
								 RBFA
							
								 LOC284240
							
								 PARD6G
							
								 LOC105376875
							
								 HSBP1L1
							
								 NFATC1
							
								 TXNL4A
							
								 LOC107985149
							
						 | 
					
				
		    		
						| nsv525694 | 
						
							
								Chr.18:79395184 - 79961125
							    on Build GRCh38
						
							
						 | 
						Loss | 
						
							
								 LOC105376875
							
								 CTDP1
							
								 LOC284241
							
								 PQLC1
							
								 KCNG2
							
								 NFATC1
							
								 LOC284240
							
						 | 
					
				
		    		
						| nsv470437 | 
						
							
								Chr.18:79657934 - 80103778
							    on Build GRCh38
						
							
						 | 
						Loss | 
						
							
								 LOC105376875
							
								 HSBP1L1
							
								 CTDP1
							
								 LOC284241
							
								 PQLC1
							
								 KCNG2
							
								 RBFADN
							
								 RBFA
							
								 TXNL4A
							
						 | 
					
				
		    		
						| nsv519406 | 
						
							
								Chr.18:79630343 - 79897319
							    on Build GRCh38
						
							
						 | 
						Gain | 
						
							
								 LOC105376875
							
								 CTDP1
							
								 LOC284241
							
								 KCNG2
							
						 | 
					
				
		    		
						| nsv1065902 | 
						
							
								Chr.18:78538045 - 80257174
							    on Build GRCh38
						
							
						 | 
						Gain | 
						
							
								 CTDP1
							
								 LOC284241
							
								 PARD6G-AS1
							
								 PQLC1
							
								 KCNG2
							
								 ADNP2
							
								 SALL3
							
								 RBFADN
							
								 ATP9B
							
								 RBFA
							
								 LOC284240
							
								 PARD6G
							
								 LOC105376875
							
								 HSBP1L1
							
								 NFATC1
							
								 LOC100996274
							
								 TXNL4A
							
								 LOC107985149
							
						 | 
					
				
		    		
						| nsv1064506 | 
						
							
								Chr.18:79580427 - 79773771
							    on Build GRCh38
						
							
						 | 
						Gain | 
						
							
								 LOC105376875
							
								 CTDP1
							
								 LOC284241
							
								 LOC284240
							
						 | 
					
				
		    		
						| nsv1066888 | 
						
							
								Chr.18:79643023 - 79769924
							    on Build GRCh38
						
							
						 | 
						Gain | 
						
							
								 LOC105376875
							
								 CTDP1
							
								 LOC284241
							
						 |