Assay Details
Target Gene Details
Entrez Gene ID: | 23210 |
Gene Name: | arginine demethylase and lysine hydroxylase |
Gene Aliases: |
PSR, PTDSR, PTDSR1 |
Location: |
Chr.17:76712830-76726799 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| JMJD6 | NM_001081461.1 | NP_001074930.1 | ||
| NM_015167.2 | NP_055982.2 | |||
| AB011157.1 | ||||
| AB073711.1 | BAC16755.1 | |||
| AK021780.1 | ||||
| AK294816.1 | ||||
| AK314541.1 | ||||
| BC047003.1 | AAH47003.1 | |||
| BC066654.1 | AAH66654.1 | |||
| DB451815.1 | ||||
| EF527406.1 | ||||
| EF527407.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833550 | Chr.17:76712038 - 76886290 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map