Assay Details
Target Gene Details
Entrez Gene ID: | 51454 |
Gene Name: | GULP, engulfment adaptor PTB domain containing 1 |
Gene Aliases: |
CED-6, CED6, GULP |
Location: |
Chr.2:188291669-188595926 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GULP1 | NM_001252668.1 | NP_001239597.1 | ||
| NM_001252669.1 | NP_001239598.1 | |||
| NM_016315.3 | NP_057399.1 | |||
| NR_045562.1 | 1 | 263 | ||
| NR_045563.1 | 1 | 263 | ||
| XM_006712580.3 | 1 | 377 | XP_006712643.1 | |
| XM_006712581.1 | XP_006712644.1 | |||
| XM_006712582.1 | XP_006712645.1 | |||
| XM_006712583.3 | 1 | 399 | XP_006712646.1 | |
| XM_006712584.3 | 1 | 399 | XP_006712647.1 | |
| XM_006712589.3 | XP_006712652.1 | |||
| XM_011511329.1 | XP_011509631.1 | |||
| XM_011511332.1 | XP_011509634.1 | |||
| XM_011511333.1 | XP_011509635.1 | |||
| XM_011511335.2 | XP_011509637.1 | |||
| XM_017004303.1 | XP_016859792.1 | |||
| XM_017004304.1 | 1 | 377 | XP_016859793.1 | |
| XM_017004305.1 | XP_016859794.1 | |||
| XM_017004307.1 | XP_016859796.1 | |||
| XM_017004308.1 | XP_016859797.1 | |||
| AF075014.1 | ||||
| AF191771.1 | 1 | 71 | AAF18975.1 | |
| AF200715.1 | 1 | 71 | AAF08006.1 | |
| AK055718.1 | ||||
| AK298626.1 | ||||
| AK314498.1 | ||||
| BC001103.2 | AAH01103.1 | |||
| CD627760.1 | 1 | 483 | ||
| DA392794.1 | 1 | 170 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv954447 | Chr.2:188290974 - 188293073 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map