Assay Details
Target Gene Details
Entrez Gene ID: | 388662 |
Gene Name: | solute carrier family 6 member 17 |
Gene Aliases: |
MRT48, NTT4 |
Location: |
Chr.1:110150510-110202202 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC6A17 | NM_001010898.2 | NP_001010898.1 | ||
| XM_006710643.2 | XP_006710706.1 | |||
| AK289982.1 | ||||
| BC140908.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv2365 | Chr.1:110168134 - 110177022 on Build GRCh38 | Deletion |
|
| nsv2354 | Chr.1:110148781 - 110180477 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map