Assay Details
Target Gene Details
Entrez Gene ID: | 10419 |
Gene Name: | protein arginine methyltransferase 5 |
Gene Aliases: |
HRMT1L5, IBP72, JBP1, SKB1, SKB1Hs |
Location: |
Chr.14:22920511-22929585 on Build GRCh38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PRMT5 | NM_001039619.2 | 12 | 1595 | NP_001034708.1 |
| NM_001282953.1 | 11 | 1463 | NP_001269882.1 | |
| NM_001282954.1 | 11 | 1476 | NP_001269883.1 | |
| NM_001282955.1 | 11 | 1378 | NP_001269884.1 | |
| NM_001282956.1 | 8 | 1133 | NP_001269885.1 | |
| NM_006109.4 | 12 | 1510 | NP_006100.2 | |
| XM_017020927.1 | 11 | 1234 | XP_016876416.1 | |
| AB451246.1 | 12 | 1286 | ||
| AB451370.1 | 12 | 1286 | ||
| AF015913.1 | 12 | 1286 | AAB66581.1 | |
| AF167572.1 | 12 | 1377 | AAF04502.1 | |
| AK075251.1 | 12 | 1388 | ||
| AK300863.1 | 11 | 1260 | ||
| AK301812.1 | 11 | 1269 | ||
| AK302240.1 | 11 | 1158 | ||
| AK315500.1 | 12 | 1326 | ||
| AL833612.1 | 12 | 1295 | ||
| BC025979.2 | 12 | 1298 | AAH25979.1 | |
| CR456741.1 | 12 | 1286 | CAG33022.1 |
More Information
Set Membership: |
|

Genomic Map