Assay Details
Target Gene Details
Entrez Gene ID: | 2314 |
Gene Name: | FLII, actin remodeling protein |
Gene Aliases: |
FLI, FLIL, Fli1 |
Location: |
Chr.17:18244815-18258916 on Build GRCh38 |
Assay Gene Location: | Within Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FLII | NM_001256264.1 | 14 | 1762 | NP_001243193.1 |
| NM_001256265.1 | 13 | 1776 | NP_001243194.1 | |
| NM_002018.3 | 14 | 1941 | NP_002009.1 | |
| XM_005256555.3 | 14 | 1730 | XP_005256612.1 | |
| XM_005256556.3 | 14 | 1727 | XP_005256613.1 | |
| XM_005256558.2 | 14 | 1727 | XP_005256615.1 | |
| AB208864.1 | 14 | 1676 | BAD92101.1 | |
| AK295655.1 | 13 | 1583 | ||
| AK295814.1 | 14 | 1844 | ||
| AK297836.1 | ||||
| AK310400.1 | 14 | 1763 | ||
| BC025300.1 | 14 | 1766 | AAH25300.1 | |
| U01184.1 | 14 | 1713 | AAC03568.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv9503 | Chr.17:18218160 - 18265852 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map