Assay Details
Target Gene Details
Entrez Gene ID: | 101927730 |
Gene Name: | uncharacterized LOC101927730 |
Gene Aliases: |
- |
Location: |
Chr.6:2883988-2900676 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC101927730 | NR_110841.1 | |||
| AY927511.1 | ||||
| AY927512.1 |
Target Gene Details
Entrez Gene ID: | 5272 |
Gene Name: | serpin family B member 9 |
Gene Aliases: |
CAP-3, CAP3, PI-9, PI9 |
Location: |
Chr.6:2887265-2903318 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SERPINB9 | NM_004155.5 | 7 | 1398 | NP_004146.1 |
| XM_005249184.4 | 7 | 1561 | XP_005249241.1 | |
| XM_011514678.2 | 7 | 1409 | XP_011512980.1 | |
| XM_017010942.1 | 8 | 1548 | XP_016866431.1 | |
| XM_017010943.1 | 8 | 1706 | XP_016866432.1 | |
| AB060690.1 | 7 | 1360 | BAB91078.1 | |
| BC002538.2 | 7 | 1359 | AAH02538.1 | |
| BX640606.1 | 2 | 876 | CAE45712.1 | |
| L40378.1 | AAC41940.1 | |||
| U71364.1 | 7 | 1389 | AAC50793.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3570730 | Chr.6:2883674 - 2890049 on Build GRCh38 | Loss |
|
| nsv830575 | Chr.6:2847376 - 3035682 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map