Assay Details
Target Gene Details
Entrez Gene ID: | 5243 |
Gene Name: | ATP binding cassette subfamily B member 1 |
Gene Aliases: |
ABC20, CD243, CLCS, GP170, MDR1, P-GP, PGY1 |
Location: |
Chr.7:87503863-87713323 on Build GRCh38 |
Assay Gene Location: | Within Exon 22 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ABCB1 | NM_000927.4 | 22 | 3092 | NP_000918.2 |
| AB208970.1 | 20 | 2562 | BAD92207.1 | |
| AF016535.1 | 21 | 2715 | AAB69423.1 | |
| AK290159.1 | 22 | 3093 | ||
| AK308467.1 | 2 | 241 | ||
| BC130424.1 | 21 | 2924 | ||
| BQ720763.1 | 3 | 267 | ||
| EU852583.1 | 20 | 2599 | ||
| EU854148.1 | 19 | 2407 | ||
| M14758.1 | 22 | 3023 | AAA59575.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv464622 | Chr.7:87203287 - 87911933 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map