Assay Details
Target Gene Details
Entrez Gene ID: | 1629 |
Gene Name: | dihydrolipoamide branched chain transacylase E2 |
Gene Aliases: |
BCATE2, BCKAD-E2, BCKADE2, BCOADC-E2, E2, E2B |
Location: |
Chr.1:100186922-100249864 on Build GRCh38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DBT | NM_001918.3 | 11 | 1615 | NP_001909.3 |
| XM_005270545.3 | 12 | 1813 | XP_005270602.1 | |
| XM_017000468.1 | 13 | 1842 | XP_016855957.1 | |
| XM_017000469.1 | 9 | 1270 | XP_016855958.1 | |
| AK303941.1 | 12 | 1786 | ||
| BC016675.1 | 11 | 1582 | AAH16675.1 | |
| J03208.1 | 11 | 1580 | AAA35589.1 | |
| M27093.1 | 11 | 1580 | AAA64512.1 | |
| X66785.1 | 11 | 1596 | CAA47285.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv830804 | Chr.1:100036777 - 100210481 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map