Assay Details
Target Gene Details
Entrez Gene ID: | 83988 |
Gene Name: | neurocalcin delta |
Gene Aliases: |
- |
Location: |
Chr.8:101686542-102124907 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 14 - Exon 14 |
More Information
Set Membership: |
|

Genomic Map