Assay Details
Target Gene Details
| Entrez Gene ID: | 132625 | 
| Gene Name: | ZFP42 zinc finger protein | 
| Gene Aliases: | REX-1, REX1, ZNF754, zfp-42 | 
| Location: | Chr.4:187994044-188005050 on Build GRCh38 | 
| Assay Gene Location: | Within Exon 5 | 
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID | 
|---|---|---|---|---|
| ZFP42 | NM_001304358.1 | 3 | 553 | NP_001291287.1 | 
| NM_174900.4 | 4 | 719 | NP_777560.2 | |
| XM_011531604.2 | 3 | 574 | XP_011529906.1 | |
| XM_011531605.1 | 4 | 724 | XP_011529907.1 | |
| XM_011531606.1 | 4 | 728 | XP_011529908.1 | |
| XM_011531607.2 | 3 | 571 | XP_011529909.1 | |
| AF450454.1 | 1 | 311 | AAL47167.1 | |
| AK056719.1 | 3 | 553 | BAB71264.1 | |
| AY429593.1 | 4 | 719 | AAR07993.1 | 
Target Copy Number Variation Details
| DGV Version: | Release date: 2016-05-15, GRCh GRCh38 | 
| Target Variation | Location | CNV Subtype | Genes | 
|---|---|---|---|
| dgv5499n100 | Chr.4:187514678 - 188139456 on Build GRCh38 | Gain |  LOC100506272  ZFP42  TRIML2  TRIML1 | 
| esv2752041 | Chr.4:187118866 - 188100311 on Build GRCh38 | Gain |  LOC100506272  ZFP42  TRIML2  LOC339975 | 
| nsv1031595 | Chr.4:187935080 - 188218104 on Build GRCh38 | Gain |  ZFP42  TRIML2  TRIML1 | 
| nsv1031097 | Chr.4:187304029 - 188382785 on Build GRCh38 | Loss |  LOC100506272  ZFP42  TRIML2  TRIML1  LOC339975 | 
| dgv9439n54 | Chr.4:186930134 - 188890842 on Build GRCh38 | Gain |  LOC100506272  LOC101930028  ZFP42  TRIML2  LINC01060  TRIML1  LOC339975 | 
| esv2763831 | Chr.4:187562041 - 189231692 on Build GRCh38 | Gain+Loss |  LOC100506272  LOC101930028  ZFP42  TRIML2  LINC01060  TRIML1 | 
| nsv596456 | Chr.4:187865684 - 188095782 on Build GRCh38 | Gain |  ZFP42  TRIML2 | 
| dgv9440n54 | Chr.4:187534043 - 188139952 on Build GRCh38 | Gain |  LOC100506272  ZFP42  TRIML2  TRIML1 | 
| nsv470098 | Chr.4:187776941 - 188300053 on Build GRCh38 | Gain |  ZFP42  TRIML2  TRIML1 | 
| nsv10629 | Chr.4:187724452 - 188143423 on Build GRCh38 | Loss |  ZFP42  TRIML2  TRIML1 | 
| dgv9441n54 | Chr.4:187540004 - 188871837 on Build GRCh38 | Loss |  LOC100506272  LOC101930028  ZFP42  TRIML2  LINC01060  TRIML1 | 
| dgv332n21 | Chr.4:187966266 - 188141060 on Build GRCh38 | Loss |  ZFP42  TRIML2  TRIML1 | 
| esv32613 | Chr.4:187056789 - 188514091 on Build GRCh38 | Gain |  LOC100506272  ZFP42  TRIML2  LINC01060  TRIML1  LOC339975 | 
| nsv1030775 | Chr.4:187983480 - 188060286 on Build GRCh38 | Loss |  ZFP42 | 
| dgv5496n100 | Chr.4:187167070 - 188594145 on Build GRCh38 | Gain |  LOC100506272  ZFP42  TRIML2  LINC01060  TRIML1  LOC339975 | 
| esv3569882 | Chr.4:187528921 - 188584766 on Build GRCh38 | Loss |  LOC100506272  ZFP42  TRIML2  LINC01060  TRIML1 | 
| nsv1027385 | Chr.4:187992714 - 188017314 on Build GRCh38 | Loss |  ZFP42 | 
| nsv830173 | Chr.4:187978318 - 188202820 on Build GRCh38 | Gain |  ZFP42  TRIML2  TRIML1 | 
| nsv1022128 | Chr.4:187957299 - 188139456 on Build GRCh38 | Loss |  ZFP42  TRIML2  TRIML1 | 
| nsv1017684 | Chr.4:187530002 - 188872654 on Build GRCh38 | Loss |  LOC100506272  LOC101930028  ZFP42  TRIML2  LINC01060  TRIML1 | 
| nsv519300 | Chr.4:188000286 - 188212954 on Build GRCh38 | Gain+Loss |  ZFP42  TRIML2  TRIML1 | 
| nsv1031236 | Chr.4:187526594 - 189035370 on Build GRCh38 | Gain |  LOC100506272  LOC101930028  ZFP42  TRIML2  LINC01060  TRIML1 | 
| nsv1022361 | Chr.4:186346140 - 188438508 on Build GRCh38 | Gain |  LOC100506272  ZFP42  F11-AS1  TRIML2  TRIML1  FAT1  LOC339975  MTNR1A | 
 Back To Top
 Back To TopMore Information
| Set Membership: |  Intragenic  Exonic  DGV Variation | 
Panther Classification:
Gene Ontology Categories:
 Back To Top
 Back To Top
 Genomic Map
Genomic Map Back To Top
 Back To Top