Assay Details
Target Gene Details
Entrez Gene ID: | 9546 |
Gene Name: | amyloid beta precursor protein binding family A member 3 |
Gene Aliases: |
MGC:15815, X11L2, mint3 |
Location: |
Chr.19:3750773-3761697 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| APBA3 | NM_004886.3 | NP_004877.1 | ||
| XM_006722950.3 | 9 | 1697 | XP_006723013.1 | |
| XM_006722951.3 | 7 | 1382 | XP_006723014.1 | |
| AB021638.1 | BAA74430.1 | |||
| AB209603.1 | 3 | 3770 | BAD92840.1 | |
| AF029110.1 | 1 | 85 | AAC17979.1 | |
| BC086306.1 | AAH86306.1 | |||
| BG938716.1 | 2 | 569 | ||
| BM977984.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv509718 | Chr.19:3676947 - 3844683 on Build GRCh38 | Insertion |
|
| nsv470114 | Chr.19:3372516 - 3855979 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map