Assay Details
Target Gene Details
Entrez Gene ID: | 1730 |
Gene Name: | diaphanous related formin 2 |
Gene Aliases: |
DIA, DIA2, DRF2, POF, POF2 |
Location: |
Chr.X:96684663-97600598 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DIAPH2 | NM_006729.4 | NP_006720.1 | ||
| NM_007309.3 | NP_009293.1 | |||
| AK291272.1 | ||||
| BC117414.1 | ||||
| BC143838.1 | ||||
| Y15908.1 | CAA75869.1 | |||
| Y15909.1 | CAA75870.1 |
Target Gene Details
Entrez Gene ID: | 29935 |
Gene Name: | replication protein A4 |
Gene Aliases: |
HSU24186 |
Location: |
Chr.X:96883908-96885467 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RPA4 | NM_013347.4 | 1 | 161 | NP_037479.1 |
| BC069791.1 | 1 | 56 | AAH69791.1 | |
| BC069808.1 | 1 | 56 | AAH69808.1 | |
| BC069824.1 | 1 | 56 | AAH69824.1 | |
| BC104013.2 | AAI04014.1 | |||
| BC104014.2 | AAI04015.1 | |||
| U24186.2 | 1 | 162 | AAB08488.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv520n21 | Chr.X:96817047 - 97148876 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map