Assay Details
Target Gene Details
Entrez Gene ID: | 64949 |
Gene Name: | mitochondrial ribosomal protein S26 |
Gene Aliases: |
C20orf193, GI008, MRP-S13, MRP-S26, MRPS13, NY-BR-87, RPMS13, dJ534B8.3 |
Location: |
Chr.20:3046029-3048250 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MRPS26 | NM_030811.3 | NP_110438.1 | ||
| AF308301.1 | AAG48268.1 | |||
| BC013018.2 | AAH13018.1 | |||
| BI115733.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1057187 | Chr.20:2906193 - 3065597 on Build GRCh38 | Gain |
|
| nsv585276 | Chr.20:3030542 - 3366307 on Build GRCh38 | Gain |
|
| nsv833897 | Chr.20:2990324 - 3175146 on Build GRCh38 | Loss |
|
| nsv833896 | Chr.20:2923851 - 3062392 on Build GRCh38 | Loss |
|
| esv33482 | Chr.20:3047082 - 3053940 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map