Assay Details
Target Gene Details
Entrez Gene ID: | 9632 |
Gene Name: | SEC24 homolog C, COPII coat complex component |
Gene Aliases: |
- |
Location: |
Chr.10:73744164-73772175 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 17 - Exon 18 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SEC24C | NM_004922.3 | NP_004913.2 | ||
| NM_198597.2 | NP_940999.1 | |||
| XM_011540380.2 | XP_011538682.1 | |||
| XM_011540381.2 | XP_011538683.1 | |||
| XM_017016967.1 | XP_016872456.1 | |||
| XM_017016968.1 | XP_016872457.1 | |||
| AK098512.1 | ||||
| AK299826.1 | ||||
| AK303085.1 | ||||
| AK309918.1 | ||||
| AK316451.1 | ||||
| AL832110.1 | ||||
| BC018928.2 | AAH18928.1 | |||
| D38555.1 | BAA07558.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1035634 | Chr.10:73724401 - 73907300 on Build GRCh38 | Gain |
|
| nsv831912 | Chr.10:73649351 - 73840445 on Build GRCh38 | Gain |
|
| nsv825468 | Chr.10:73715689 - 73922563 on Build GRCh38 | Gain |
|
| nsv551516 | Chr.10:73749515 - 73847410 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map