Assay Details
Target Gene Details
Entrez Gene ID: | 328 |
Gene Name: | apurinic/apyrimidinic endodeoxyribonuclease 1 |
Gene Aliases: |
APE, APE1, APEN, APEX, APX, HAP1, REF1 |
Location: |
Chr.14:20455131-20457772 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| APEX1 | NM_001244249.1 | NP_001231178.1 | ||
| NM_001641.3 | NP_001632.2 | |||
| NM_080648.2 | NP_542379.1 | |||
| NM_080649.2 | NP_542380.1 | |||
| AK098588.1 | ||||
| AK291100.1 | ||||
| AK311548.1 | ||||
| BC002338.2 | AAH02338.1 | |||
| BC004979.1 | AAH04979.1 | |||
| BC008145.1 | AAH08145.1 | |||
| BC019291.1 | AAH19291.1 | |||
| BC095428.1 | AAH95428.1 | |||
| BT007236.1 | AAP35900.1 | |||
| BT020133.1 | AAV38935.1 | |||
| BX161454.1 | CAD61917.1 | |||
| CB139337.1 | ||||
| D90373.1 | BAA14381.1 | |||
| M80261.1 | AAA58371.1 | |||
| M81955.1 | AAA58372.1 | |||
| S43127.1 | AAB22977.1 | |||
| U79268.1 | AAB50212.1 | |||
| X59764.1 | CAA42437.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv23438 | Chr.14:20332606 - 20789719 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map