Assay Details
Target Gene Details
Entrez Gene ID: | 4524 |
Gene Name: | methylenetetrahydrofolate reductase (NAD(P)H) |
Gene Aliases: |
- |
Location: |
Chr.1:11785730-11806103 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MTHFR | NM_005957.4 | 2 | 301 | NP_005948.3 |
| XM_005263458.3 | 2 | 1138 | XP_005263515.1 | |
| XM_005263460.4 | 2 | 756 | XP_005263517.1 | |
| XM_005263462.4 | 2 | 189 | XP_005263519.1 | |
| XM_005263463.3 | 2 | 301 | XP_005263520.1 | |
| XM_011541495.2 | 2 | 1135 | XP_011539797.1 | |
| XM_011541496.2 | 2 | 1138 | XP_011539798.1 | |
| XM_017001328.1 | 2 | 1138 | XP_016856817.1 | |
| AB209113.1 | 1 | 339 | BAD92350.1 | |
| AJ237672.1 | 1 | 201 | CAB41971.1 | |
| AK312907.1 | 2 | 162 | ||
| AY046560.1 | AAL17646.1 | |||
| AY046561.1 | AAL17647.1 | |||
| AY046562.1 | AAL17648.1 | |||
| AY046563.1 | AAL17649.1 | |||
| AY046564.1 | AAL17650.1 | |||
| AY046565.1 | AAL17651.1 | |||
| BC053509.1 | 2 | 123 | AAH53509.1 |
More Information
Set Membership: |
|

Genomic Map