Assay Details
Target Gene Details
Entrez Gene ID: | 201161 |
Gene Name: | centromere protein V |
Gene Aliases: |
3110013H01Rik, CENP-V, PRR6, p30 |
Location: |
Chr.17:16342534-16353498 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CENPV | NM_181716.2 | 1 | 422 | NP_859067.2 |
| AF514992.1 | 1 | 358 | AAM76703.1 | |
| BC052604.1 | 1 | 423 | AAH52604.1 | |
| BC137486.1 | 1 | 454 | ||
| BC137487.1 | 1 | 454 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1063764 | Chr.17:16268335 - 16759600 on Build GRCh38 | Loss |
|
| nsv827903 | Chr.17:16348665 - 16380446 on Build GRCh38 | Gain |
|
| esv2715702 | Chr.17:15750514 - 16668268 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map