Assay Details
Target Gene Details
Entrez Gene ID: | 7132 |
Gene Name: | TNF receptor superfamily member 1A |
Gene Aliases: |
CD120a, FPF, MS5, TBP1, TNF-R, TNF-R-I, TNF-R55, TNFAR, TNFR1, TNFR1-d2, TNFR55, TNFR60, p55, p55-R, p60 |
Location: |
Chr.12:6328757-6342117 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TNFRSF1A | NM_001065.3 | 10 | 1515 | NP_001056.1 |
| A21522.1 | 10 | 1366 | CAA01558.1 | |
| AK056611.1 | 10 | 1414 | ||
| AK130807.1 | 3 | 1225 | ||
| AK291088.1 | 10 | 1441 | ||
| AK298729.1 | 9 | 1248 | ||
| AK304517.1 | 9 | 1209 | ||
| AK308061.1 | 10 | 1295 | ||
| AK315509.1 | 10 | 1512 | ||
| BC010140.2 | 10 | 1443 | AAH10140.1 | |
| JN172914.1 | 9 | 1339 | ||
| M33294.1 | 10 | 1393 | AAA03210.1 | |
| M58286.1 | 10 | 1398 | AAA36753.1 | |
| M60275.1 | 10 | 1366 | AAA36756.1 | |
| M63121.1 | 10 | 1418 | AAA36754.1 |
More Information
Set Membership: |
|

Genomic Map