Assay Details
Target Gene Details
Entrez Gene ID: | 56912 |
Gene Name: | intraflagellar transport 46 |
Gene Aliases: |
C11orf2, C11orf60, CFAP32 |
Location: |
Chr.11:118544528-118575624 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 10 - Intron 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| IFT46 | NM_001168618.1 | NP_001162089.1 | ||
| NM_020153.3 | NP_064538.3 | |||
| XM_011542905.2 | XP_011541207.1 | |||
| XM_011542906.2 | XP_011541208.1 | |||
| XM_017018017.1 | XP_016873506.1 | |||
| XM_017018018.1 | XP_016873507.1 | |||
| AJ249980.1 | CAB96537.1 | |||
| AK025480.1 | BAB15146.1 | |||
| AK289521.1 | ||||
| AL136934.1 | CAB66868.1 | |||
| BC011647.2 | AAH11647.1 | |||
| BC012802.2 | AAH12802.1 | |||
| BC022856.1 | AAH22856.1 | |||
| CR457344.1 | CAG33625.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1044281 | Chr.11:118503814 - 118554704 on Build GRCh38 | Gain |
|
| esv3579929 | Chr.11:118544686 - 118568073 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map