Assay Details
Target Gene Details
Entrez Gene ID: | 64902 |
Gene Name: | alanine--glyoxylate aminotransferase 2 |
Gene Aliases: |
AGT2, DAIBAT |
Location: |
Chr.5:34998101-35048188 on Build GRCh38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| AGXT2 | NM_001306173.1 | 10 | 1275 | NP_001293102.1 |
| NM_031900.3 | 12 | 1500 | NP_114106.1 | |
| XM_005248337.2 | 12 | 1550 | XP_005248394.1 | |
| XM_005248338.2 | 10 | 1064 | XP_005248395.1 | |
| XM_017009748.1 | 10 | 1034 | XP_016865237.1 | |
| AB193309.1 | 12 | 1287 | BAD66662.1 | |
| AJ292204.1 | 12 | 1259 | CAC24841.1 | |
| AK055732.1 | 4 | 927 | ||
| AK055757.1 | 12 | 1129 | ||
| AK223128.1 | 12 | 1222 | BAD96848.1 | |
| AK223144.1 | 12 | 1248 | ||
| AK223375.1 | 12 | 1314 | BAD97095.1 | |
| AK314691.1 | 12 | 1314 | ||
| BC144268.1 | 10 | 1084 | ||
| BC150603.1 | 10 | 1084 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv597810 | Chr.5:34948427 - 35060217 on Build GRCh38 | Gain |
|
| nsv1028968 | Chr.5:34945017 - 35060217 on Build GRCh38 | Gain |
|
| esv3604691 | Chr.5:34934667 - 35064045 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map