Assay Details
Target Gene Details
Entrez Gene ID: | 51397 |
Gene Name: | COMM domain containing 10 |
Gene Aliases: |
PTD002 |
Location: |
Chr.5:116084592-116446462 on Build GRCh38 |
Assay Gene Location: | Within Intron 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| COMMD10 | XM_017009557.1 | XP_016865046.1 |
Target Gene Details
Entrez Gene ID: | 57556 |
Gene Name: | semaphorin 6A |
Gene Aliases: |
HT018, SEMA, SEMA6A1, SEMAQ, VIA |
Location: |
Chr.5:116443555-116574926 on Build GRCh38 |
Assay Gene Location: | Within Exon 23 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SEMA6A | NM_001300780.1 | 20 | 5949 | NP_001287709.1 |
| NM_020796.4 | 19 | 5898 | NP_065847.1 | |
| XM_006714663.1 | 20 | 5863 | XP_006714726.1 | |
| XM_017009675.1 | 20 | 5268 | XP_016865164.1 | |
| XM_017009676.1 | 15 | 7815 | XP_016865165.1 | |
| AF225425.1 | 1 | 998 | AAG09727.1 | |
| AK027505.1 | 1 | 965 | ||
| AK027867.1 | 17 | 5025 | BAB55418.1 |
More Information
Set Membership: |
|

Genomic Map