Assay Details
Target Gene Details
Entrez Gene ID: | 23258 |
Gene Name: | DENN domain containing 5A |
Gene Aliases: |
RAB6IP1 |
Location: |
Chr.11:9138825-9265335 on Build GRCh38 |
Assay Gene Location: | Within Exon 26 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DENND5A | NM_001243254.1 | 23 | 4176 | NP_001230183.1 |
| NM_015213.3 | 23 | 4210 | NP_056028.2 | |
| XM_005252832.1 | 24 | 4279 | XP_005252889.1 | |
| XM_011519953.2 | 19 | 2533 | XP_011518255.1 | |
| AB029014.1 | 23 | 4170 | ||
| AK025499.1 | 18 | 2821 | BAB15155.1 | |
| AK026007.1 | 16 | 2284 | ||
| AK091148.1 | 5 | 1854 | ||
| AK125444.1 | 15 | 3515 | ||
| AK295881.1 | 23 | 3937 | ||
| AK302478.1 | 20 | 3671 | ||
| AL117448.1 | 23 | 3966 | CAB55932.2 | |
| BC009354.2 | 19 | 2938 | AAH09354.2 | |
| BC071596.1 | 24 | 4015 | AAH71596.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv33264 | Chr.11:8986065 - 9289584 on Build GRCh38 | Loss |
|
| nsv526778 | Chr.11:9138192 - 9163530 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map