Assay Details
Target Gene Details
Entrez Gene ID: | 51156 |
Gene Name: | serpin family A member 10 |
Gene Aliases: |
PZI, ZPI |
Location: |
Chr.14:94280460-94293353 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SERPINA10 | NM_001100607.2 | NP_001094077.1 | ||
| NM_016186.2 | NP_057270.1 | |||
| XM_005267733.4 | XP_005267790.1 | |||
| XM_017021353.1 | XP_016876842.1 | |||
| AF181467.1 | AAD53962.1 | |||
| AK314877.1 | ||||
| AY358597.1 | AAQ88960.1 | |||
| BC022261.1 | AAH22261.1 | |||
| BX248011.1 | CAD62339.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832862 | Chr.14:94163327 - 94374084 on Build GRCh38 | Gain |
|
| esv3635377 | Chr.14:94176964 - 94375000 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map